missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trometamol, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Trometamol
Specifications
Specifications
| Chemical Name or Material | Trometamol |
| CAS | 77-86-1 |
| Analyte or Component Names | Trometamol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C4 H11 N O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Trometamol, aminomethylidynetri(methanol), 1,3-Propanediol, 2-amino-2-(hydroxymethyl)-, 2-Amino-1,3-dihydroxy-2-(hydroxymethyl)propane, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-hydroxymethylpropan-1,3-diol, 2-Amino-2-methylol-1,3-propanediol, Addex-Tham, Aminotri(hydroxymethyl)methane, Aminotrimethylolmethane, Aminotris(hydroxymethyl)methane, Methanamine, 1,1,1-tris(hydroxymethyl)-, NSC 103026, NSC 6365, NSC 65434, Pehanorm, TAM, TAM (buffering agent), THAM, Talatrol, Tri Amino, Tri(hydroxymethyl)aminomethane, Tri(hydroxymethyl)methylamine, Trigmo base, Triladyl, Trimethylolaminomethane, Tris, Tris (buffering agent), Tris Amino, Tris Amino Crystal, Tris base, Tris buffer, Tris(hydroxymethyl)aminomethane, Tris(hydroxymethyl)methanamine, Tris(hydroxymethyl)methylamine, Tris(methylolamino)methane, Tris-steril, Trisamin, Trisamine, Trisaminol, Trizma, Trizma Base, Trometamole, Tromethamine, Tromethane, Tromethanmin, Tutofusin tris, [2-Hydroxy-1,1-bis(hydroxymethyl)ethyl]amine |
| InChI Formula | InChI=1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2 |
| SMILES | NC(CO)(CO)CO |
| IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction