missing translation for 'onlineSavingsMsg'
Learn More

Trolox, TRC

Product Code. 17993585
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17993585 10 g 10g
17983495 1 g 1g
17923405 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17993585 Supplier Toronto Research Chemicals Supplier No. TRCT89256010G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53188-07-1
Molecular Formula C14 H18 O4
Molecular Weight (g/mol) 250.2903
Synonym 3,4-Dihydro-6-hydroxy-2,5,7,8-tetramethyl-2H-1-benzopyran-2-carboxylic Acid, (±)-3,4-Dihydro-6-hydroxy-2,5,7,8-tetramethyl-2H-1-Benzopyran-2-carboxylic Acid, (R,S)-6-Hydroxy-2,5,7,8-tetramethyl-2-chromanecarboxylic Acid, (±)-6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic acid, (±)-Trolox, 6-Hydroxy-2,5,7,8-tetramethyl-3,4-dihydro-2H-chromene-2-carboxylic Acid, 6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic Acid, Trolox C
IUPAC Name 6-hydroxy-2,5,7,8-tetramethyl-3,4-dihydrochromene-2-carboxylic acid
SMILES CC1=C(C2=C(C(C)=C1O)CCC(C)(C(O)=O)O2)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C14H18O4/c1-7-8(2)12-10(9(3)11(7)15)5-6-14(4,18-12)13(16)17/h15H,5-6H2,1-4H3,(H,16,17)
Formula Weight 250.1205
Percent Purity >95%
Chemical Name or Material Trolox
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.