missing translation for 'onlineSavingsMsg'
Learn More

Trolox™, 97%

Product Code. 10002910 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10002910 25 g Glass bottle 25g
10454155 1 g Glass bottle 1g
10782831 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10002910 Supplier Thermo Scientific Acros Supplier No. 218940250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 53188-07-1 | C14H18O4 | 250.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53188-07-1
Molecular Formula C14H18O4
Molecular Weight (g/mol) 250.29
MDL Number MFCD00006846
InChI Key GLEVLJDDWXEYCO-UHFFFAOYNA-N
Synonym trolox, 6-hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic acid, trolox c, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl-2h-1-benzopyran-2-carboxylic acid, 6-hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 2h-1-benzopyran-2-carboxylic acid, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl, +/--6-hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 2h-1-benzopyran-2-carboxylic acid, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl-, +-, trolox™, trolox tm
PubChem CID 40634
ChEBI CHEBI:82625
IUPAC Name 6-hydroxy-2,5,7,8-tetramethyl-3,4-dihydrochromene-2-carboxylic acid
SMILES CC1=C(C)C2=C(CCC(C)(O2)C(O)=O)C(C)=C1O

Specifications

Melting Point 187.0°C to 194.0°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Beilstein 18, V,7, 335
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in methanol, ethanol
Formula Weight 250.29
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Trolox*, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Trolox
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Trolox

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.