Learn More
LTE Scientific™ Trolley Touchclave Loading/Carriage
Product Code. 12785385
Change view
Click to view available options
For Use With (Equipment):
150 L Model Autoclaves
200 L Model Autoclaves
300 L Model Autoclaves
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12785385 300 L Model Autoclaves Each
12765385 150 L Model Autoclaves Each
12775385 200 L Model Autoclaves -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12785385 Supplier LTE Scientific™ Supplier No. TC/LCT/03
  • ONLINE EXCLUSIVE PRICE
    £2935.00
    £2880.00 / Pack of 1
    Save £55.00
    1% Off
Estimated Shipment: 20-04-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Loading trolley for LTE autoclaves provides easy manoeuvrability of loads to and from the autoclave. Reduces manual handling for staff safety.
  • Loading trolley/carriage for LTE Autoclaves
  • Reduced manual handling
  • Easy and secure manoeuvrability of loads
Specifications
Type Loading Trolley
Holds Equipment requiring sterilization
Material Stainless Steel
For Use With (Equipment) 300 L Model Autoclaves