missing translation for 'onlineSavingsMsg'
Learn More

Troleandomycin, TRC

Product Code. 30009863
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
2.5 mg
Unit Size:
0.5mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009863 2.5 mg 2.5mg
17911375 0.5 mg 0.5mg
17950775 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009863 Supplier Toronto Research Chemicals Supplier No. TRCT8925332.5MG
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2751-09-9
Molecular Formula C41H67NO15
Molecular Weight (g/mol) 813.97
Synonym Oleandomycin, 2' ',4',11-triacetate, Oleandomycin triacetate (6CI), Oleandomycin, triacetate (ester) (8CI,9CI), Oxiraneheptanoic acid, 2-(3,5-dihydroxy-2,4-dimethylhexanoyl)-δ-[[4-(dimethylamino)tetrahydro-3-hydroxy-6-methylpyran-2-yl]oxy]-α,γ,ε-trimethyl-β-[(tetrahydro-5-hydroxy-4-methoxy-6-methylpyran-2-yl)oxy]-, μ-lactone, triacetate (7CI), 1,9-Dioxaspiro[2.13]hexadecane, oleandomycin deriv., Acetyloleandomycin, Cyclamycin, Evramicina, Evramycin, NSC 108166, Oleandomycin triacetyl ester, Oleomycin, TAO, TAO (pharmaceutical), Triacetyloleandomycin, Triocetin, Troleandomycin, Wy 651, Wytrion
IUPAC Name [(3R,5S,6S,7R,8S,9R,12R,13S,14S,15R)-6-[(2S,3R,4S,6R)-3-acetyloxy-4-(dimethylamino)-6-methyloxan-2-yl]oxy-8-[(2R,4S,5S,6S)-5-acetyloxy-4-methoxy-6-methyloxan-2-yl]oxy-5,7,9,12,13,15-hexamethyl-10,16-dioxo-1,11-dioxaspiro[2.13]hexadecan-14-yl] acetate
SMILES CO[C@H]1C[C@H](O[C@H]2[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3OC(=O)C)N(C)C)[C@@H](C)C[C@@]4(CO4)C(=O)[C@H](C)[C@@H](OC(=O)C)[C@@H](C)[C@@H](C)OC(=O)[C@@H]2C)O[C@@H](C)[C@@H]1OC(=O)C

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C41H67NO15/c1-19-17-41(18-49-41)38(46)23(5)34(53-27(9)43)21(3)25(7)52-39(47)24(6)35(56-32-16-31(48-14)36(26(8)51-32)54-28(10)44)22(4)33(19)57-40-37(55-29(11)45)30(42(12)13)15-20(2)50-40/h19-26,30-37,40H,15-18H2,1-14H3/t19-,20+,21-,22+,23+,24+,25+,26-,30-,31-,32-,33-,34-,35-,36-,37+,40-,41+/m0/s1
Formula Weight 813.4511
Percent Purity >95%
Chemical Name or Material Oleandomycin triacetate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.