missing translation for 'onlineSavingsMsg'
Learn More

Troglitazone, MedChemExpress

Product Code. 16474057
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16474057 50 mg 50mg
16468148 10 mg 10mg
16417906 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16474057 Supplier Medchem Express Supplier No. HY50935
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Troglitazone is a PPARγ agonist, with EC50s of 550 nM and 780 nM for human and murine PPARγ receptor, respectively.

Troglitazone is a PPARγ agonist, with EC50s of 550 nM and 780 nM for human and murine PPARγ receptor, respectively.

Specifications

Chemical Name or Material Troglitazone
CAS 97322-87-7
Color White
Purity Grade Notes Research
Molecular Formula C24H27NO5S
Quantity 50 mg
Synonym CS-045
Solubility Information DMSO : 160 mg/mL (362.37 mM; Need ultrasonic)
SMILES O=C(SC1CC(C=C2)=CC=C2OCC3(C)OC(C(CC3)=C4C)=C(C)C(C)=C4O)NC1=O
Molecular Weight (g/mol) 441.54
Formula Weight 441.54
Percent Purity 96.46%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.