missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TROAP Antibody [Alexa Fluor™ 700], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-45721AF700
This item is not returnable.
View return policy
Description
TROAP Polyclonal antibody specifically detects TROAP in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| TROAP | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 10024 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Alexa Fluor 700 | |
| TASTIN, trophinin assisting protein, trophinin associated protein (tastin), Trophinin-assisting protein, trophinin-assisting protein (tastin), Trophinin-associated protein | |
| trophinin associated protein(tastin). (Uniprot# Q12815) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction