missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TRO 19622

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Neuroprotective and neuroregenerative compound. Rescues motor neurons from axotomy-induced cell death and promotes nerve regeneration following sciatic nerve crush in vivo. Binds directly to two components of the mitochondrial permeability pore, the voltage-dependent anion channel (VDAC) and translocator protein; thought to inhibit MPTP opening.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 399.65 |
| Purity | >98% |
| Molecular Weight (g/mol) | 399.663 |
| Chemical Name or Material | TRO 19622 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction