missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRNT1 Polyclonal Antibody

Description
Peptide sequence: PQDIDFATTA TPTQMKEMFQ SAGIRMINNR GEKHGTITAR LHEENFEITT Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 93%; Zebrafish: 93%.
TRNT1 belongs to the tRNA nucleotidyltransferase/poly(A) polymerase family. It adds and repairs the conserved 3'-CCA sequence necessary for the attachment of amino acids to the 3' terminus of tRNA molecules, using CTP and ATP as substrates.
Specifications
Specifications
| Antigen | TRNT1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | TRNT1 |
| Gene Accession No. | Q96Q11 |
| Gene Alias | 2410043H24Rik; 2610044E04Rik; 9830143O18Rik; AA408384; ATP(CTP):tRNA nucleotidyltransferase; bMtCCA; C76540; CCA tRNA nucleotidyltransferase 1, mitochondrial; CCA1; CCA-adding enzyme; CGI-47; mitochondrial CCA-adding tRNA-nucleotidyltransferase; mitochondrial tRNA nucleotidyl transferase, CCA-adding; mt CCA-adding enzyme; mt tRNA adenylyltransferase; mt tRNA CCA-diphosphorylase; mt tRNA CCA-pyrophosphorylase; MtCCA; mt-Trat; RPEM; SIFD; tRNA adenylyltransferase, mitochondrial; tRNA CCA nucleotidyl transferase 1; tRNA nucleotidyl transferase 1; tRNA nucleotidyl transferase, CCA-adding, 1; tRNA nucleotidyl transferase-like; tRNA-nucleotidyltransferase; tRNA-nucleotidyltransferase 1 mitochondrial-like protein; tRNA-nucleotidyltransferase 1, mitochondrial; TRNT1; Unknown (protein for MGC:140399); zgc:86645 |
| Gene Symbols | TRNT1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?