missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRNT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557878
This item is not returnable.
View return policy
Description
Immunogen sequence: LDLRLKIAKE EKNLGLFIVK NRKDLIKATD SSDPLKPYQD FIIDSREPDA TTRVCELLKY QGEHCLLKEM QQWSIPPFPV SGHDIR Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
TRNT1 belongs to the tRNA nucleotidyltransferase/poly(A) polymerase family. It adds and repairs the conserved 3'-CCA sequence necessary for the attachment of amino acids to the 3' terminus of tRNA molecules, using CTP and ATP as substrates.
Specifications
| TRNT1 | |
| Polyclonal | |
| Unconjugated | |
| TRNT1 | |
| 2410043H24Rik; 2610044E04Rik; 9830143O18Rik; AA408384; ATP(CTP):tRNA nucleotidyltransferase; bMtCCA; C76540; CCA tRNA nucleotidyltransferase 1, mitochondrial; CCA1; CCA-adding enzyme; CGI-47; mitochondrial CCA-adding tRNA-nucleotidyltransferase; mitochondrial tRNA nucleotidyl transferase, CCA-adding; mt CCA-adding enzyme; mt tRNA adenylyltransferase; mt tRNA CCA-diphosphorylase; mt tRNA CCA-pyrophosphorylase; MtCCA; mt-Trat; RPEM; SIFD; tRNA adenylyltransferase, mitochondrial; tRNA CCA nucleotidyl transferase 1; tRNA nucleotidyl transferase 1; tRNA nucleotidyl transferase, CCA-adding, 1; tRNA nucleotidyl transferase-like; tRNA-nucleotidyltransferase; tRNA-nucleotidyltransferase 1 mitochondrial-like protein; tRNA-nucleotidyltransferase 1, mitochondrial; TRNT1; Unknown (protein for MGC:140399); zgc:86645 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 312616, 51095 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96Q11 | |
| TRNT1 | |
| Recombinant protein corresponding to Human TRNT1. Recombinant protein control fragment (Product #RP-96279). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction