Learn More
TRNT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NBP1-86589
This item is not returnable.
View return policy
Description
TRNT1 Polyclonal specifically detects TRNT1 in Human, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications
| Western Blot, Immunofluorescence | |
| Western Blot 0.04 to 0.4 μg/mL, Simple Western 1:8, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200-1:500, Knockdown Validated | |
| 50 kDa | |
| IgG |
| Polyclonal | |
| TRNT1 | |
| Human, Rat |