Learn More
Invitrogen™ TRMT6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114089
Description
Immunogen sequence: GAMMERMGGF GSIIQLYPGG GPVRAATACF GFPKSFLSGL YEFPLNKVDS For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
Substrate-binding subunit of tRNA (adenine-N(1)-)-methyltransferase, which catalyzes the formation of N(1)-methyladenine at position 58 (m1A58) in initiator methionyl-tRNA.
Specifications
| TRMT6 | |
| Polyclonal | |
| Unconjugated | |
| TRMT6 | |
| 3300001M20Rik; AU016412; AW492787; CGI-09; GCD10; Gcd10p; Kiaa1153; MGC5029; mKIAA1153; mRNA methyladenosine-N(1)-methyltransferase non-catalytic subunit TRM6; RGD1308877; Trm6; TRMT6; tRNA (adenine(58)-N(1))-methyltransferase non-catalytic subunit TRM6; tRNA (adenine-N(1)-)-methyltransferase non-catalytic subunit TRM6; tRNA methyltransferase 6; tRNA methyltransferase 6 homolog; tRNA(m1A58)-methyltransferase subunit TRM6; tRNA(m1A58)MTase subunit TRM6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51605 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UJA5 | |
| TRMT6 | |
| Synthetic peptide directed towards the middle region of human TRMT6. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.