missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRMT6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565616
This item is not returnable.
View return policy
Description
Immunogen sequence: LKDKGIKGEEI VQQLIENSTT FRDKTEFAQD KYIKKKKKKY EAIITVVKPS TRILSIMYYA REPGKINHMR YDTLAQMLTL GNIRAGN Highest antigen sequence identity to the following orthologs - mouse 97%, rat 98%.
Substrate-binding subunit of tRNA (adenine-N(1)-)-methyltransferase, which catalyzes the formation of N(1)-methyladenine at position 58 (m1A58) in initiator methionyl-tRNA.
Specifications
| TRMT6 | |
| Polyclonal | |
| Unconjugated | |
| TRMT6 | |
| 3300001M20Rik; AU016412; AW492787; CGI-09; GCD10; Gcd10p; Kiaa1153; MGC5029; mKIAA1153; mRNA methyladenosine-N(1)-methyltransferase non-catalytic subunit TRM6; RGD1308877; Trm6; TRMT6; tRNA (adenine(58)-N(1))-methyltransferase non-catalytic subunit TRM6; tRNA (adenine-N(1)-)-methyltransferase non-catalytic subunit TRM6; tRNA methyltransferase 6; tRNA methyltransferase 6 homolog; tRNA(m1A58)-methyltransferase subunit TRM6; tRNA(m1A58)MTase subunit TRM6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51605 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UJA5 | |
| TRMT6 | |
| Recombinant Human TRMT6. Recombinant protein control fragment (Product #RP-106283). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction