missing translation for 'onlineSavingsMsg'
Learn More

TRMT5 Antibody (OTI1B5) [HRP], Novus Biologicals™

Product Code. 30688122 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30688122 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688122 Supplier Novus Biologicals Supplier No. NBP344935H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRMT5 Monoclonal antibody specifically detects TRMT5 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRMT5
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone OTI1B5
Conjugate HRP
Formulation PBS
Gene Alias KIAA1393EC 2.1.1.31, M1G-methyltransferase, TRM5 tRNA methyltransferase 5 homolog (S. cerevisiae), TRM5MGC111453, tRNA (guanine-N(1)-)-methyltransferase, tRNA [GM37] methyltransferase, tRNA methyltransferase 5, tRNA-N1G37 methyltransferase
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-303 of human TRMT5(NP_065861) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57570
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.