missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRMT2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557161
This item is not returnable.
View return policy
Description
Immunogen sequence: FYLGTWRDGN VVCVQSNHLK NIPEKHSQVA QYYEVFLRQS PLEPCLVFHE GGYWRELTVR TNSQGHTMAI ITFHPQKLSQ EELHVQKEIV KE Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 32%.
Probable S-adenosyl-L-methionine-dependent methyltransferase that catalyzes the formation of 5-methyl-uridine at position 54 (m5U54) in all tRNA. May also have a role in tRNA stabilization or maturation.
Specifications
| TRMT2B | |
| Polyclonal | |
| Unconjugated | |
| TRMT2B | |
| CXorf34; dJ341D10.3; TRM2 homolog; TRM2 tRNA methyltransferase 2 homolog B; TRMT2B; tRNA (uracil(54)-C(5))-methyltransferase homolog; tRNA (uracil-5-)-methyltransferase homolog; tRNA methyltransferase 2 homolog B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79979 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96GJ1 | |
| TRMT2B | |
| Recombinant protein corresponding to Human TRMT2B. Recombinant protein control fragment (Product #RP-95473). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction