missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRMT2A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA596575
This item is not returnable.
View return policy
Description
Immunogen sequence: LASQHLVAIL DPPRAGLHSK VILAIRRAKN LRRLLYVSCN PRAAMGNFVD LCRAPSNRVK GIPFRPVKAV AVDLFPQTPH CEMLILFERV EHPNGTGVLG PHSPPAQPTP GPPDNTLQET GTFPSS Positive Samples: HL-60, Raji, NCI-H460
The protein encoded by TRMT2A is of unknown function. However, it is orthologous to the mouse Trmt2a gene and contains an RNA methyltransferase domain. Expression of TRMT2A varies during the cell cycle, with aberrant expression being a possible biomarker in certain breast cancers. Several transcript variants encoding two different isoforms have been found for TRMT2A.
Specifications
| TRMT2A | |
| Polyclonal | |
| Unconjugated | |
| TRMT2A | |
| HpaII tiny fragments locus 9c, hpaII tiny fragments locus 9c protein, Htf9c, Htf9-c, TRM2 tRNA methyltransferase 2 homolog A, TRM2 tRNA methyltransferase 2A, TRMT2A, tRNA (uracil-5-)-methyltransferase homolog A, tRNA methyltransferase 2 homolog A, tRNA methyltransferase 2 homolog A (S. cerevisiae) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 27037 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide, pH 7.3 | |
| Q8IZ69 | |
| TRMT2A | |
| A synthetic peptide corresponding to a sequence within amino acids 500 to the C-terminus of human TRMT2A/HTF9C (NP_0735643) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction