missing translation for 'onlineSavingsMsg'
Learn More

TRMT12 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18433256 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433256 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433256 Supplier Novus Biologicals Supplier No. NBP176583JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRMT12 Polyclonal antibody specifically detects TRMT12 in Human, Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRMT12
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias Alpha-amino-alpha-carboxypropyl transferase TYW2, EC 2.1.1, EC 2.1.1.32, EC 2.5.1.-, FLJ20772, homolog of yeast tRNA methyltransferase, Trm12, tRNA methyltranferase 12 homolog, tRNA methyltranferase 12 homolog (S. cerevisiae), tRNA methyltransferase 12 homolog (S. cerevisiae), tRNA-yW synthesizing protein 2, tRNA-yW-synthesizing protein 2, TYW2tRNA wybutosine-synthesizing protein 2 homolog
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human TYW2. The immunogen is located within the first 50 amino acids of TYW2. Amino Acid Squence: TQRYREYLQRQKLFDTQHR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 55039
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.