Learn More
TRMT112 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26472-1-AP-150UL
This item is not returnable.
View return policy
Description
The TRMT112 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TRMT112. This antibody recognizes human antigen. The TRMT112 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| TRMT112 | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| HSPC152, HSPC170, hypothetical protein HSPC152, TRM112 like protein, TRMT112 | |
| 150 μL | |
| 51504 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |