missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRMT112 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558971
This item is not returnable.
View return policy
Description
Immunogen sequence: MKLLTHNLLS SHVRGVGSRG FPLRLQATEV RICPVEFNPN FVARMIPKVE WSAFLEAADN LRL Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
Specifications
| TRMT112 | |
| Polyclonal | |
| Unconjugated | |
| TRMT112 | |
| AD-001; HSPC152; HSPC170; hTrm112; Multifunctional methyltransferase subunit TRM112-like protein; TRM112; TRM112-like protein; TRMT112; TRMT11-2; tRNA methyltransferase 112 homolog; tRNA methyltransferase 11-2 homolog (S. cerevisiae) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51504 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UI30 | |
| TRMT112 | |
| Recombinant protein corresponding to Human TRMT112. Recombinant protein control fragment (Product #RP-97477). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction