Learn More
TrkC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527729
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, Molt-4, Raji, NIH-3T3. Predicted reactivity: Mouse (91%), Rat (91%), Pig (96%), Chicken (81%), Rhesus Monkey (100%), Chimpanzee (99%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NTRK3 (TRKC) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system. Known substrates for the TRK receptors are SHC, PI-3 kinase, and PLCG1. The different isoforms do not have identical signaling properties. The protein is widely expressed, mainly in the nervous tissue. The isoform B is expressed in a relatively large amount in the adult brain comparatively to fetal brain. TRKC is subject to ligand-mediated auto-phosphorylation. The protein structure contains 2 immunoglobulin-like C2-type domains and 2 leucine-rich (LRR) repeats.
Specifications
| TrkC | |
| Polyclonal | |
| Unconjugated | |
| Ntrk3 | |
| AW125844; ETS related protein-neurotrophic receptor tyrosine kinase fusion protein; ETV6-NTRK3 fusion; gp145(trkC); GP145-TrkC; neural receptor protein-tyrosine kinase (trkC); neurotrophic receptor tyrosine kinase 3; Neurotrophic tyrosine kinase receptor type 3; neurotrophic tyrosine kinase, receptor, type 3; neurotrophin-3; NT-3 growth factor receptor; NTRK3; Ntrk3_tv3; TrkC; trk-C; trkC tyrosine kinase; tyrosine kinase receptor; tyrosine kinase receptor C | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18213, 4916 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q16288, Q6VNS1 | |
| Ntrk3 | |
| Recombinant fragment corresponding to a region within amino acids 222 and 468 of Human TrkC | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.