missing translation for 'onlineSavingsMsg'
Learn More

TrkB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen TrkB
Concentration 600 μg/mL
Applications Immunohistochemistry, Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17721129
View Documents
Proteintech
29961-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17711129
View Documents
Proteintech
29961-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TrkB Polyclonal antibody specifically detects TrkB in Human, Mouse, Rat samples. It is validated for IHC, WB, ELISA
Specifications

Specifications

TrkB
Immunohistochemistry, Western Blot, ELISA
Unconjugated
Rabbit
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
IgG
Antigen affinity purification
Antibody
600 μg/mL
Polyclonal
Liquid
Human, Mouse, Rat
4915
Primary
Store at -20°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.