missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TrkB Antibody (Rinat patent anti-TrkB) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28744JF669
This item is not returnable.
View return policy
Description
TrkB Monoclonal antibody specifically detects TrkB in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| TrkB | |
| Monoclonal | |
| Janelia Fluor 669 | |
| BDNF/NT-3 growth factors receptor, EC 2.7.10, EC 2.7.10.1, GP145-TrkB, Neurotrophic tyrosine kinase receptor type 2, neurotrophic tyrosine kinase, receptor, type 2, trk-B, TrkB tyrosine kinase, TRKBBDNF-tropomyosine receptor kinase B, tyrosine kinase receptor B | |
| TrkB/ NTRK2 | |
| 0.1 mL | |
| Neuroendocrinology, Oncogenes, Phospho Specific, Protein Kinase, Signal Transduction | |
| 4915 | |
| Store at 4°C in the dark. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| Rinat patent anti-TrkB | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction