missing translation for 'onlineSavingsMsg'
Learn More

TrkB Antibody (OTI2E1), DyLight 594, Novus Biologicals™

Product Code. 18418236 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418236 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418236 Supplier Novus Biologicals Supplier No. NBP274624DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TrkB Monoclonal antibody specifically detects TrkB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TrkB
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2E1
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias BDNF/NT-3 growth factors receptor, EC 2.7.10, EC 2.7.10.1, GP145-TrkB, Neurotrophic tyrosine kinase receptor type 2, neurotrophic tyrosine kinase, receptor, type 2, trk-B, TrkB tyrosine kinase, TRKBBDNF-tropomyosine receptor kinase B, tyrosine kinase receptor B
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human TrkB expression vector.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroendocrinology, Oncogenes, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4915
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.