missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TrkA/TrkB/TrkC Recombinant Rabbit Monoclonal Antibody (RM423)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA536218
This item is not returnable.
View return policy
Description
MA5-36218 reacts to human TRK. This antibody may also react to mouse or rat TRK, as predicted by immunogen homology.
Specifications
| TrkA/TrkB/TrkC | |
| Recombinant Monoclonal | |
| 0.08 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
| P04629, Q16288, Q16620 | |
| NTRK1, Ntrk2, NTRK3 | |
| A peptide corresponding to the C-terminus of human TRK. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| RM423 | |
| Unconjugated | |
| NTRK1 | |
| BDNF/NT-3 growth factors receptor; BDNF-tropomyosine receptor kinase B; C80751; gp140trk; GP145-TrkB; GP145-TrkB/GP95-TrkB; high affinity nerve growth factor receptor; MTC; neural receptor protein-tyrosine kinase (trkB); neurotrophic receptor tyrosine kinase 1; neurotrophic receptor tyrosine kinase 2; Neurotrophic tyrosine kinase receptor type 1; Neurotrophic tyrosine kinase receptor type 2; neurotrophic tyrosine kinase, receptor, type 1; neurotrophic tyrosine kinase, receptor, type 2; neurotrophic tyrosine receptor kinase type 2; Ntrk1; NTRK2; Oncogene TRK; p140-TrkA; RATTRKB1; Slow nerve growth factor receptor; Tkr; Tkrb; trk; TRK1; TRK1-transforming tyrosine kinase protein; TRKA; trk-A; TrkA neurotrophin receptor; trkA proto-oncogene receptor; TRKB; trk-B; TrkB tyrosine kinase; TRKB1; tropomyosin-related kinase A; Tropomyosin-related kinase B; Tyrosine kinase receptor; Tyrosine kinase receptor A; tyrosine kinase receptor B | |
| Rabbit | |
| Protein A | |
| RUO | |
| 4914, 4915, 4916 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction