missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX TrkA Protein, Active

Product Code. 17340364
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17340364

Brand: Eurofins DiscoverX 14571

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, N-terminal His6-tagged recombinant human TrkA, residues 440-end, for use in Kinase Assays.

Three tyrosine kinase receptors, designated TRKA, TRKB and TRKC, have been identified that selectively bind NGF, BDNF and NT-3, respectively; NT-4/5 also binds to the TRKB receptor. NGF binds to their specific TRK receptor to stimulate intracellular signaling pathways leading to specific protein phosphorylations and differential gene inductions. TRKA (High affinity nerve growth factor, TRK1 transforming tyrosine kinase protein, p140-TrkA, Trk-A) receptor kinases engage the activation of a branched proto-oncoprotein signaling pathway involving Shc, Src, Ras, Raf, mitogen-activated protein kinase, and other downstream effectors. Inhibitors of TRKA may be useful in the treatment of cancer.

GenBank NM_002529

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 8.5 (at 4°C), 300 mM NaCl, 5% glycerol, 0.1 mM EGTA, 0.03% Brij-35, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 41 kDa
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal 6His
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Truncated
Purity or Quality Grade ≥66%
Protein TRKA, active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.