Learn More
Invitrogen™ TrkA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120786
Description
Positive test controls include: HeLa, K-562, NIH/3T3, Mouse liver, Mouse heart, Rat liver. The target is usually found in the following locations: Cell membrane, Early endosome membrane, Late endosome membrane, Single-pass type I membrane protein. Immunogen sequence: MLRGGRRGQL GWHSWAAGPG SLLAWLILAS AGAAPCPDAC CPHGSSGLRC TRDGALDSLH HLPGAENLTE LYIENQQHLQ HLELRDLRGL GELRNLTIVK SGLRFVAPDA FHFTPRLSRL NLSFNALESL SWKTVQGLSL QELVLSGNPL HCSCALRWLQ RWEEEGLGGV PEQKLQCHGQ GPLAHMPNAS CGVPTLKVQV PNASVDVGDD VLLRCQVEGR GLEQAGWILT ELEQSATVMK SGGLPSLGLT LANVTSDLNR KNVTCWAEND VGRAEVSVQV.
NTRK1 (TRKA) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system. TrkA is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. The presence of TrkA leads to cell differentiation and may play a role in specifying sensory neuron subtypes. Mutations in TrkA have been associated with congenital insensitivity to pain, anhidrosis, self-mutilating behavior, mental retardation and cancer.
Specifications
| TrkA | |
| Polyclonal | |
| Unconjugated | |
| Ntrk1 | |
| C80751; DKFZp781I14186; EC 2.7.10.1; gp140trk; high affinity nerve growth factor receptor; kinase TrkA; MTC; neurotrophic receptor tyrosine kinase 1; Neurotrophic tyrosine kinase receptor type 1; neurotrophic tyrosine kinase, receptor, type 1; Ntrk1; Oncogene TRK; p140-TrkA; Slow nerve growth factor receptor; Tkr; TRK; TRK1; TRK1-transforming tyrosine kinase protein; Trka; Trk-A; TrkA neurotrophin receptor; trkA proto-oncogene receptor; tropomyosin-related kinase A; Tyrosine kinase receptor; Tyrosine kinase receptor A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18211, 4914, 59109 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P04629, P35739, Q3UFB7 | |
| Ntrk1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human TrkA (NP_002520.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.