missing translation for 'onlineSavingsMsg'
Learn More

TrkA/B Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18321817 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18321817 20 μg 20µL
18327954 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18321817 Supplier Novus Biologicals Supplier No. NBP31689720UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TrkA/B Polyclonal antibody specifically detects TrkA/B in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TrkA/B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias BDNF/NT-3 growth factors receptor, DEE58, EIEE58, gp140trk, GP145-TrkB, High affinity nerve growth factor receptor, MTC, Neurotrophic tyrosine kinase receptor type 1, 2, NTRK1, NTRK2, OBHD, p140-TrkA, TRK1-transforming tyrosine kinase protein, Tropomyosin-related kinase A, B, Tyrosine kinase receptor A, B
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around Y674 & Y675 of human TrkA (NP_001012331.1).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 4914
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.