missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trk (pan) Antibody (SR1160), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21696-100ul
This item is not returnable.
View return policy
Description
Trk (pan) Monoclonal antibody specifically detects Trk (pan) in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
| Trk (pan) | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 4914 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| SR1160 | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| High affinity nerve growth factor receptor;MTC;NTRK1;Nucleoprotein TPR;p140-TrkA;Protein TFG;TRK;TRK1;TRKA;Trk-A;Tyrosine-protein kinase receptor | |
| A synthesized peptide derived from human Trk (pan) (Uniprot #: P04629, Q16288, Q16620) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction