missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRK fused gene Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565710
This item is not returnable.
View return policy
Description
Immunogen sequence: LRRELIELRNK VNRLLDSLEP PGEPGPSTNI PENDTVDGRE EKSASDSSGK QSTQVMAASM SAFDPLKNQD EINKNVMSAF GLTDDQVSGP PS Highest antigen sequence identity to the following orthologs - mouse 96%, rat 97%.
TFG protein, a recently identified novel protein, is a human homolog of mouse TFG protein. It contains an N-terminal coiled-coil structure, a SPYGQ-rich region followed by octicosapeptide repeats. Research shows that TFG can modulate SHP-1 activity and is a novel member of the NF-kB pathway. The physiological functions of this protein remain largely elusive, but TFG is thought to function by associating with other proteins, including TANK and NEMO. TFG was first identified as a partner of NTRK1 and is involved in generating the thyroid TRK-T3 oncogene, as well as oncogenic rearrangements with ALK in anaplastic lymphoma and NOR1 in mixoid chondrosarcoma.
Specifications
| TRK fused gene | |
| Polyclonal | |
| Unconjugated | |
| TFG | |
| AI173908; HMSNP; protein TFG; SPG57; TF6; TFG; Trk-fused gene; TRK-fused gene protein; TRKT3; TRKT3 oncogene | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10342 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q92734 | |
| TFG | |
| Recombinant Human TFG. Recombinant protein control fragment (Product #RP-104940). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction