missing translation for 'onlineSavingsMsg'
Learn More

Trizma™ base, TraceSELECT™, ≥99.9995% (Metals basis), Honeywell™ Fluka™

Product Code. 15615500 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15615500 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15615500 Supplier Honeywell-Fluka Supplier No. 42616100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

TraceSELECT™, for inorganic trace analysis,≥99.9995% (metals basis)

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

TRUSTED_SUSTAINABILITY

Specifications

CAS 77-86-1
Melting Point 170°C
Boiling Point 288°C
Color White
pH 10.2 to 10.6 (at 6.00 g/L 20°C)
Physical Form Solid
Quantity 100 g
Molecular Formula C4H11NO3
Linear Formula NH2C(CH2OH)3
Odor Odorless
MDL Number MFCD00004679
Packaging Poly bottle
UN Number NONH for all modes of transport
Beilstein 741883
Solubility Information 100.0g/L at 20°C in water
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
SMILES C(C(CO)(CO)N)O
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular Weight (g/mol) 121.136
PubChem CID 6503
ChEBI CHEBI:9754
Formula Weight 121.14g/mol
Percent Purity ≥99.9995% (metals basis)
Grade TraceSELECT
Chemical Name or Material Trizma™ base
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.