missing translation for 'onlineSavingsMsg'
Learn More

Tritylamine, 98%

Product Code. 11493890
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493890 1 g 1g
11421538 5 g 5g
11431538 25 g 25g
11441538 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11493890 Supplier Thermo Scientific Alfa Aesar Supplier No. A13864.03
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5824-40-8 | C19H17N | 259.352 g/mol

Triphenylmethylamine was used to prepare diamondoid porous organic salt.1 It was also used in preparation of N-tritylated β-amino alcohols, useful building blocks in organic synthesis. It is used in the preparation of trityl- cysteine analogs as inhibitors of human mitotic kinesin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylmethylamine was used to prepare diamondoid porous organic salt.1 It was also used in preparation of N-tritylated β-amino alcohols, useful building blocks in organic synthesis. It is used in the preparation of trityl- cysteine analogs as inhibitors of human mitotic kinesin.

Solubility
Soluble in chloroform, methanol. Insoluble in water.

Notes
Air sensitive. Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5824-40-8
Molecular Formula C19H17N
Molecular Weight (g/mol) 259.352
MDL Number MFCD00008047
InChI Key BZVJOYBTLHNRDW-UHFFFAOYSA-N
Synonym tritylamine, triphenylmethylamine, benzenemethanamine, .alpha.,.alpha.-diphenyl, tritylamin, trityl amine, aminotriphenylmethane, triphenylaminomethane, triphenyl methylamine, aminotrityl, polymer-bound
PubChem CID 138598
IUPAC Name triphenylmethanamine
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N

Specifications

Melting Point 100°C to 104°C
Boiling Point 223°C (14 mmHg)
Quantity 1 g
Beilstein 2113674
Solubility Information Soluble in chloroform,methanol. Insoluble in water.
Formula Weight 259.35
Percent Purity 98%
Chemical Name or Material Tritylamine
compliance-icons
Product Identifier
  • Tritylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Tritylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.