missing translation for 'onlineSavingsMsg'
Learn More

Trityl Chloride, TRC

Product Code. 30451499
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451499 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451499 Supplier Toronto Research Chemicals Supplier No. TRCT887260250G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trityl Chloride
CAS 76-83-5
Molecular Formula C19H15Cl
Quantity 250 g
Synonym 1,1',1''-(Chloromethylidyne)tris[benzene], Chlorotriphenylmethane, NSC 435, Triphenylchloromethane, Triphenylmethyl Chloride, Tr-Cl, TrCl
InChI Formula InChI=1S/C19H15Cl/c20-19(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H
SMILES ClC(c1ccccc1)(c2ccccc2)c3ccccc3
IUPAC Name [chloro(diphenyl)methyl]benzene
Molecular Weight (g/mol) 278.78
Formula Weight 278.0862
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.