missing translation for 'onlineSavingsMsg'
Learn More

J.T. Baker™ Triton™ X100

Product Code. 10421871 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10421871 500 mL 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10421871 Supplier J.T. Baker™ Supplier No. 2838.0500

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

BAKER grade laboratory reagent, packaged in a glass bottle.

BAKER grade chemicals are for laboratory use, for organic synthesis, and for routine research usually controlled by physical criteria and often by assay (by GC).

Specifications

Color Pale Yellow
Includes Glass bottle
Molecular Formula (C2H4O)nC14H22O
Formula Weight 647g/mol
Synonym Octylphenoxy polyethoxy ethanol, TX-100, Mono-300
Solubility Soluble
Content And Storage Store in cool, dry place
Form Liquid
For Use With (Application) Detergent, cell lysis
Quantity 500 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Triton X100
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Polyethylene glycoloctylphenyl ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.