missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Triton™ X-405
Brand: Thermo Scientific Chemicals 215690025
View more versions of this product
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Triton™ X-405 | |
70% Solution in Water | |
Liquid | |
-4.0°C | |
2.5 L | |
Glass bottle | |
1.08 | |
MFCD00132505 | |
Solubility in water: soluble. | |
CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1 | |
250.38 | |
1968.47 |
Yellow | |
9002-93-1, 7732-18-5 | |
120.0°C | |
1.0800g/mL | |
>109°C | |
1.4360 to 1.4380 | |
C16H26O2 | |
15, 6850 | |
JYCQQPHGFMYQCF-UHFFFAOYSA-N | |
2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol | |
5590 | |
≥65% |
Safety and Handling
Product Identifier
Hazard Category
Hazard Statement
Precautionary Statement
- Triton]TS X-405, 70% solution in water
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
RUO – Research Use Only