missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Triton™ X-114 is used in biochemical applications to solubilize and separate proteins due to its low cloud point (23°C).
- At temperatures above the cloud point, detergent solutions separate into aqueous and detergent-enriched phases.
- This makes Triton™ X-114 particularly useful in separating lipophilic proteins from hydrophilic proteins.
Specifications
Specifications
| Color | Light Yellow |
| Physical Form | Liquid |
| Density | 8.8 lb./gal |
| Quantity | 500 mL |
| Packaging | Plastic Container |
| Specific Gravity | 1.054 g/ml |
| Molecular Formula | (C2H4O)x · C14H22O (x= 7-8) |
| Molecular Weight (g/mol) | 294.435 g/mol |
| Chemical Name or Material | Triton™ X-114 |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?