missing translation for 'onlineSavingsMsg'
Learn More

Triton™ X-100, Molecular Biology Reagent Grade, MP Biomedicals™

Product Code. 11446631 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
Unit Size:
100mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446631 100 mL 100mL
11436631 50 mL 50mL
11456631 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11446631 Supplier MP Biomedicals Supplier No. 0219485480

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
Synonym 4-(1,1,3,3-Tetramethylbutyl)phenyl-polyethylene glycol, 4-(1, 1, 3
PubChem CID 5590
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Specifications

Color Colorless to Yellow
Physical Form Viscous Liquid
Density 8.9 lb./gal.
Quantity 100 mL
Grade Molecular Biology
Chemical Name or Material Triton™ X-100
compliance-icons compliance-icons compliance-icons
Product Identifier
  • tirton X-100, molecular biology reagent
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: triton X-100
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.