missing translation for 'onlineSavingsMsg'
Learn More

Triton™ X-100 (Electrophoresis), Fisher BioReagents™

Product Code. 10254583
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass Bottle
HDPE Bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10254583 500 mL HDPE Bottle 500mL
10102913 100 mL Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10254583 Supplier Fisher BioReagents Supplier No. BP151500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

For solubilization and permeabilization of membranes

This nonionic detergent is used to solubilize membranes, permeabilize unfixed eukaryotic membranes, and is a component of lysis buffers for DNA extraction.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
Synonym Polyethylene Glycol p-tert-Octylphenyl Ether
PubChem CID 5590
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Specifications

Color Undesignated
Physical Form Liquid
Boiling Point 270°C
Melting Point 6°C
Cloud Point 63 to 69°C
Density 1.067g/cm³
pH 6 to 8
Quantity 500 mL
Identification Pass Test
Packaging HDPE Bottle
Linear Formula 4-(C8H17) C6H4 (OCH2CH2)n OH (n∽10)
Merck Index 15, 6850
Solubility Information Soluble in water
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol
Formula Weight 646.86
Grade Electrophoresis
Chemical Name or Material Triton™ X-100
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Triton X-100]TS
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.