missing translation for 'onlineSavingsMsg'
Learn More

Triton X-100

Product Code. 11408706
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408706 2500 mL 2500mL
11488696 100 mL 100mL
11498696 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11408706 Supplier Thermo Scientific Alfa Aesar Supplier No. A16046.0F
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 9002-93-1 | C16H26O2 | 250.38 g/mol

Detergent, equivalent to Nonidet P-40.Triton™ X-100 is used as a non-ionic detergent, emulsifier and dispersing agent. It is useful for isolation of membrane proteins. It is also used in electrophoresis. It is an active ingredient in influenza vaccine (Fluzone). It finds applications in the dispersion of carbon materials for soft composite materials and restricting colony expansion in Aspergillus nidulans in microbiology. Further, it is an active ingredient in homemade vinyl record cleaning fluids together with distilled water and isopropyl alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Detergent, equivalent to Nonidet P-40.Triton|r X-100 is used as a non-ionic detergent, emulsifier and dispersing agent. It is useful for isolation of membrane proteins. It is also used in electrophoresis. It is an active ingredient in influenza vaccine (Fluzone). It finds applications in the dispersion of carbon materials for soft composite materials and restricting colony expansion in Aspergillus nidulans in microbiology. Further, it is an active ingredient in homemade vinyl record cleaning fluids together with distilled water and isopropyl alcohol.

Solubility
Miscible with water.

Notes
Store in a cool place. Incompatible with strong acids, strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
PubChem CID 5590
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Specifications

Boiling Point >250°C
Melting Point ∽5°C
Density 1.070
Quantity 2500 mL
Flash Point 180°C (356°F)
Refractive Index 1.49
Solubility Information Miscible with water.
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol
Formula Weight ca. 625
Chemical Name or Material Triton X-100
compliance-icons
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.