missing translation for 'onlineSavingsMsg'
Learn More

Tritolyl phosphate, 63-85%, technical, residue other tolyl phosphate isomers

Product Code. 10721715 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
2.5kg
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10721715 2.5 kg Glass bottle 2.5kg
10055690 25 g Glass bottle 25g
10687391 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10721715 Supplier Thermo Scientific Acros Supplier No. 422310025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 1330-78-5 | C21H21O4P | 368.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-78-5
Molecular Formula C21H21O4P
Molecular Weight (g/mol) 368.36
MDL Number MFCD02102116
Synonym Tricresyl phosphate

Specifications

Melting Point -30°C
Color White
Density 1.1600g/mL
Boiling Point 265°C (10.0 mmHg)
Flash Point 234°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (sum of isomers) (GC)
Packaging Glass bottle
Linear Formula (CH3C6H4O)3PO
Refractive Index 1.5550 to 1.557
Quantity 2.5 kg
Merck Index 15, 9940
Specific Gravity 1.16
Solubility Information Solubility in water: insoluble
Formula Weight 368.36
Percent Purity 99%
Grade Technical
Physical Form Powder
Chemical Name or Material Tritolyl phosphate, tech., 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tritolyl phosphate, mixture of isomers
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P391-Collect spillage.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.