missing translation for 'onlineSavingsMsg'
Learn More

Tritolyl phosphate, 63-85%, technical, residue other tolyl phosphate isomers

Product Code. 10055690 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
2.5kg
25g
500g
This item is not returnable. View return policy

Product Code. 10055690

Brand: Thermo Scientific Acros 422310250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1330-78-5 | C21H21O4P | 368.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1330-78-5
Molecular Formula C21H21O4P
Molecular Weight (g/mol) 368.36
MDL Number MFCD02102116
Synonym Tricresyl phosphate

Spezifikation

Melting Point -30°C
Color White
Density 1.1600g/mL
Boiling Point 265°C (10.0 mmHg)
Flash Point 234°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (sum of isomers) (GC)
Packaging Glass bottle
Linear Formula (CH3C6H4O)3PO
Refractive Index 1.5550 to 1.557
Quantity 25 g
Merck Index 15, 9940
Specific Gravity 1.16
Solubility Information Solubility in water: insoluble
Formula Weight 368.36
Percent Purity 99%
Grade Technical
Physical Form Powder
Chemical Name or Material Tritolyl phosphate, tech., 99%
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • Tritolyl phosphate, mixture of isomers
Signalwort
  • Warning
Gefahrenkategorie
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 2
Gefahrenhinweis
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
Sicherheitshinweis
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P391-Collect spillage.

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado