missing translation for 'onlineSavingsMsg'
Learn More

Tribenzylamine, 99+%

Product Code. p-202373 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Unit Size:
100g
1kg
250g
Product Code. Quantity unitSize
10071810 100 g 100g
10276200 250 g 250g
10239443 1 kg 1kg
3 options
This item is not returnable. View return policy

Product Code. 10071810

Brand: Thermo Scientific Acros 156471000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 620-40-6 | C21H21N | 287.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-40-6
Molecular Formula C21H21N
Molecular Weight (g/mol) 287.41
MDL Number MFCD00004773
InChI Key MXHTZQSKTCCMFG-UHFFFAOYSA-N
Synonym tribenzylamine, benzenemethanamine, n,n-bis phenylmethyl, unii-hz10o1931j, trisbenzylamine, n,n-dibenzyl-1-phenyl-methanamine, pubchem21076, acmc-1atok, dsstox_cid_27031, dsstox_rid_82052
PubChem CID 24321
IUPAC Name N,N-dibenzyl-1-phenylmethanamine
SMILES C(N(CC1=CC=CC=C1)CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 90°C to 94°C
Color White to Cream
Boiling Point 380°C to 390°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula (C6H5CH2)3N
Quantity 100 g
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in ethanol,soluble in toluene
Formula Weight 287.39
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material Tribenzylamine, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Tribenzylamine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.