missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Biosciences™ Tritest: CD3 FITC/CD8 PE/CD45 PerCP (CE/IVD)
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
Description
- Clone SK7 (also known as Leu-4) SK1 2D1 (CE/IVD)
- This BD Tritest™ CD3 fluorescein isothiocyanate (FITC)/CD8 phycoerythrin (PE)/CD45 peridinin chlorophyll protein (PerCP) is a three-color direct immunofluorescence reagent for use with a suitably equipped flow cytometer to identify and determine the percentages and absolute counts of mature human T lymphocytes (CD3+) and suppressor/cytotoxic (CD3+CD8+) T-lymphocytes subsets in erythrocyte-lysed whole blood.
- When used with BD Trucount™ tubes, absolute counts of these populations can be enumerated from a single tube.
- This BD Tritest reagent and Trucount tubes can be used with the FACS™ Loader. The reagent can be used with or without an isotype control.
- Contents:
- Specificity: CD3, Clone: SK7 (Leu4), Format: FITC, Isotype: IgG<sub>1</sub> K, Entrez Gen ID: 916
- Specificity: CD8, Clone: SK1, Format: PE, Isotype: IgG<sub>1</sub> K, Entrez Gen ID: 925
- Specificity: CD45, Clone: 2D1, Format: PerCP, Isotype: IgG<sub>1</sub> K
Specifications
Specifications
| Content And Storage | Store the reagent at 2°C–8°C to maintain stability. Do not use after expiration date shown on the label. Do not freeze the reagent or expose it to direct light during storage or during incubation with cells. Keep the reagent vial dry. Do not use the reagent if you observe any changes in appearance. Precipitation or discoloration indicates instability or deterioration. |
| No. of Tests | 50 Tests |
| Quantity | 50 Tests |
| Regulatory Status | CE/IVD |
| Product Type | Immunofluorescence Flow Cytometry Test Reagents |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction