missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Biosciences™ Tritest: CD3 FITC/CD19 PE/CD45 PerCP (CE/IVD)
Click to view available options
Quantity:
50 Tests
Unit Size:
1 set
Description
- Clone SK7 (Leu-4) SJ25C1 (SJ25-C2) 2D1 (CE/IVD)
- BD TriTEST™ CD3 fluorescein isothiocyanate (FITC)/CD19 phycoerythrin (PE)/CD45 peridinin chlorophyll protein (PerCP) is a three-color direct immunofluorescence reagent for use with a suitably equipped flow cytometer to identify and determine the percentages and absolute counts of mature human T (CD3+) and B (CD19+) lymphocytes in erythrocytelysed whole blood.
- When used with TruCOUNT™ Tubes, absolute counts of these populations can be enumerated from a single tube.
- BD TriTEST reagent and TruCOUNT Tubes can be used with the FACS Loader. The reagent can be used with or without an isotype control.
- Contents:
- Specificity: CD3, Clone: SK7 (also known as Leu4), Format FITC, Isotype IgG<sub>1</sub> K, Entrez Gene ID: 916
- Specificity: CD19, Clone SJ25C1, Format: PE, Isotype: IgG<sub>1</sub> K, Entrez Gene ID: 930
- Specificity: CD45, Clone: 2D1, Format: PerCP, Isotype: IgG<sub>1</sub> K
Specifications
Specifications
| Content And Storage | Store the reagent at 2°C–8°C to maintain stability. Do not use after expiration date shown on the label. Do not freeze the reagent or expose it to direct light during storage or during incubation with cells. Keep the reagent vial dry. Do not use the reagent if you observe any changes in appearance. Precipitation or discoloration indicates instability or deterioration. |
| No. of Tests | 50 Tests |
| Quantity | 50 Tests |
| Regulatory Status | CE/IVD |
| Product Type | Immunofluorescence Flow Cytometry Test Reagents |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction