missing translation for 'onlineSavingsMsg'
Learn More

BD Biosciences™ Tritest: CD3 FITC/CD16+CD56 PE/CD45 PerCP with BD Trucount™ Tubes (CE/IVD)

Product Code. 15895998
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15895998 50 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15895998 Supplier BD Biosciences™ Supplier No. 342442

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Three-color direct immunofluorescence reagent for determining percentages and absolute counts of mature human T lymphocytes (CD3+), and natural killer (NK) (CD3-, CD16+, CD56+) lymphocytes in erythrocyte-lysed whole blood.

  • Clone SK7 (Leu-4) B73.1 NCAM16.2 (NCAM 16) 2D1 (CE/IVD)
  • BD Tritest™ CD3 fluorescein isothiocyanate (FITC)/CD16 + CD56 phycoerythrin (PE)/CD45 peridinin chlorophyll protein (PerCP) is a threecolor direct immunofluorescence reagent for use with a suitably equipped flow cytometer to identify and determine the percentages and absolute counts of mature human T lymphocytes (CD3+) and natural killer (NK) (CD3CD16+CD56+) lymphocyte subsets in erythrocyte-lysed whole blood.
  • When used with BD Trucount™ tubes, absolute counts of these populations can be enumerated from a single tube.
  • BD Tritest reagent and BD Trucount tubes can be used with the BD FACS™ Loader. The reagent can be used with or without an isotype control.
  • Contents:
  • Specificity: CD3, Clone: SK7 (also known as Leu4), Format FITC, Isotype IgG<sub>1</sub> K, Entrez Gen ID: 916
  • Specificity: CD16 (FcyRIII), Clone: B73.1, Format: PE, Isotype IgG<sub>1</sub> K
  • Specificity: CD56 (NCAM-1) Clone: NCAM16.2 )NCAM 16), Format: PE, Isotype: IgG<sub>2b</sub> K, Entrez Gene ID: 4684
  • Specificity: CD45, Clone: 2D1, Format: PerCP, Isotype: IgG<sub>1</sub> K
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the reagent at 2°C–8°C to maintain stability. Do not use after expiration date shown on the label. Do not freeze the reagent or expose it to direct light during storage or during incubation with cells. Keep the reagent vial dry. Do not use the reagent if you observe any changes in appearance. Precipitation or discoloration indicates instability or deterioration.
No. of Tests 50 Tests
Quantity 50 Tests
Regulatory Status CE/IVD
Product Type Immunofluorescence Flow Cytometry Test Reagents
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.