missing translation for 'onlineSavingsMsg'
Learn More

Trisphenyl(3,3,3-trifluoroprop-1-yl)phosphonium Iodide, TRC

Product Code. 30452107
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452107 50 mg 50mg
30450740 100 mg -
30454015 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30452107 Supplier Toronto Research Chemicals Supplier No. TRCT89629550MG
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128622-15-1
Molecular Formula C21H19F3P.I
Molecular Weight (g/mol) 486.2
Synonym Phosphonium, triphenyl(3,3,3-trifluoropropyl)-, iodide (1:1) (ACI), Phosphonium, triphenyl(3,3,3-trifluoropropyl)-, iodide (9CI), (3,3,3-Trifluoropropyl)triphenylphosphonium iodide, Triphenyl(3,3,3-trifluoropropyl)phosphonium iodide, Trisphenyl(3,3,3-trifluoroprop-1-yl)phosphonium iodide
IUPAC Name triphenyl(3,3,3-trifluoropropyl)phosphanium;iodide
SMILES [I-].FC(F)(F)CC[P+](c1ccccc1)(c2ccccc2)c3ccccc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C21H19F3P.HI/c22-21(23,24)16-17-25(18-10-4-1-5-11-18,19-12-6-2-7-13-19)20-14-8-3-9-15-20;/h1-15H,16-17H2;1H/q+1;/p-1
Formula Weight 486.0221
Chemical Name or Material Trisphenyl(3,3,3-trifluoroprop-1-yl)phosphonium Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.