missing translation for 'onlineSavingsMsg'
Learn More

Tris, Ultra Pure grade, 99.95% min., MP Biomedicals™

Product Code. 11493541 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
10 kg
Unit Size:
10kg
1kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493541 1 kg 1kg
11403551 5 kg 5kg
11413551 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493541 Supplier MP Biomedicals Supplier No. 0210313301

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym 2-Amino-2-(hydroxymethyl)-1, 3-propanediol, Tris base
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range >99.9%
Formula Weight 121.1
Grade Ultrapure
Chemical Name or Material Tris
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.