missing translation for 'onlineSavingsMsg'
Learn More

Tris(triphenylphosphine)ruthenium(II) chloride, 98%

Product Code. 10197893 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10197893 25 g Glass bottle 25g
10541651 1 g Glass bottle 1g
10124550 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10197893 Supplier Thermo Scientific Acros Supplier No. 207880250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15529-49-4 | C54H45Cl2P3Ru | 958.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15529-49-4
Molecular Formula C54H45Cl2P3Ru
Molecular Weight (g/mol) 958.85
MDL Number MFCD00013077
InChI Key KSHWMFNJCNGTJO-UHFFFAOYSA-L
Synonym dichlorotris triphenylphosphine ruthenium ii, tris triphenylphosphine ruthenium ii chloride, dichlorotris triphenylphosphino ruthenium ii, tris triphenylphosphine ruthenium ii dichloride, dichlorotris triphenylphosphine ruthenium, ruthenium ii-tris triphenylphosphine dichloride, tris triphenylphosphine dichlororuthenium, ruthenium dichlorotris triphenylphosphine, pubchem12822
PubChem CID 11007548
IUPAC Name dichlororuthenium;triphenylphosphane
SMILES [Cl-].[Cl-].[Ru+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 159.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula [(C6H5)3P]3RuCl2
Quantity 25 g
Fieser 04,564; 05,740; 06,654; 12,179; 16,126; 06,175; 07,101; 08,840; 09,104; 17,33
Solubility Information Solubility in water: insoluble. Other solubilities: sparingly soluble in methanol, ethanol, acetone,, ethyl acetate, chloroform and toluene, insoluble in diethyl ether and n-hexane
Formula Weight 958.85
Percent Purity 98%
Physical Form Solution
Chemical Name or Material Tris(triphenylphosphine)ruthenium(II) chloride
Show More Show Less
compliance-icons
Product Identifier
  • Tris(triphenylphosphine)ruthenium(II) chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.