missing translation for 'onlineSavingsMsg'
Learn More

Tris(triphenylphosphine)rhodium(I) chloride, 99.99%, (trace metal basis)

Product Code. 10309483 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10309483 250 mg Glass bottle 250mg
10646212 1 g Glass bottle 1g
10741661 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10309483 Supplier Thermo Scientific Supplier No. 194572500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 14694-95-2 | C54H45ClP3Rh | 925.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14694-95-2
Molecular Formula C54H45ClP3Rh
Molecular Weight (g/mol) 925.23
MDL Number MFCD00010016
InChI Key IXAYKDDZKIZSPV-UHFFFAOYSA-M
Synonym chlorotris triphenylphosphine rhodium i, chlorotris triphenylphosphine rhodium, tris triphenylphosphine rhodium i chloride, tris triphenylphosphine chlororhodium, wilkinson's catalyst, unii-0fv534bkev, tris triphenylphosphine rhodium chloride, 0fv534bkev, rhodium; triphenylphosphane; chloride
PubChem CID 84599
SMILES [Cl-].[Rh+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 245.0°C to 250.0°C
Color Maroon
Infrared Spectrum Authentic
Assay Percent Range 99.99%
Packaging Glass bottle
Linear Formula [(C6H5)3P]3RhCl
Quantity 250 mg
Fieser 01,140; 02,448; 03,325; 04,559; 05,736; 06,652; 07,68; 08,109; 09,113; 15,90
Merck Index 15, 10245
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform, dichloromethane, benzene,, toluene, less soluble in acetic acid, ketones, methanol and, lower aliphatic alcohols, virtually insoluble in alkanes and cyclohexane
IUPAC Name tris(triphenylphosphane) hydrochloridyl rhodium
Formula Weight 925.23
Percent Purity 99.99%
Grade Trace Metal Basis
Physical Form Crystalline Powder
Chemical Name or Material Tris(triphenylphosphine)rhodium(I) chloride, 99.99%
Show More Show Less
compliance-icons
Product Identifier
  • Tris(triphenylphosphine)rhodium(I) chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Tris(triphenylphosphine)rhodium(I) chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.