missing translation for 'onlineSavingsMsg'
Learn More

Tris(triphenylphosphine)rhodium(I) chloride, 98%

Product Code. 10104260 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10104260 5 g Glass bottle 5g
10793191 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10104260 Supplier Thermo Scientific Acros Supplier No. 190420050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14694-95-2 | C54H45ClP3Rh | 925.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14694-95-2
Molecular Formula C54H45ClP3Rh
Molecular Weight (g/mol) 925.23
MDL Number MFCD00010016
InChI Key IXAYKDDZKIZSPV-UHFFFAOYSA-M
Synonym chlorotris triphenylphosphine rhodium i, chlorotris triphenylphosphine rhodium, tris triphenylphosphine rhodium i chloride, tris triphenylphosphine chlororhodium, wilkinson's catalyst, unii-0fv534bkev, tris triphenylphosphine rhodium chloride, 0fv534bkev, rhodium; triphenylphosphane; chloride
PubChem CID 84599
SMILES [Cl-].[Rh+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 245.0°C to 250.0°C
Color Brown-Red to Maroon
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula [(C6H5)3P]3RhCl
Quantity 5 g
Fieser 01,140; 02,448; 03,325; 04,559; 05,736; 06,652; 07,68; 08,109; 09,113; 15,90
Merck Index 15, 10245
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform, dichloromethane, benzene,, toluene, less soluble in acetic acid, ketones, methanol and, lower aliphatic alcohols, virtually insoluble in alkanes and cyclohexane
IUPAC Name tris(triphenylphosphane) hydrochloridyl rhodium
Formula Weight 925.23
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Tris(triphenylphosphine)rhodium(I) chloride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.