missing translation for 'onlineSavingsMsg'
Learn More

Tris(trimethylsilyl) borate

Product Code. 10340194 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10340194 100 g Glass bottle 100g
10539203 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10340194 Supplier Thermo Scientific Acros Supplier No. 381241000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4325-85-3 | C9H27BO3Si3 | 278.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4325-85-3
Molecular Formula C9H27BO3Si3
Molecular Weight (g/mol) 278.38
MDL Number MFCD00051588
InChI Key YZYKZHPNRDIPFA-UHFFFAOYSA-N
Synonym tris trimethylsilyl borate, tris trimethylsiloxy boron, silanol, trimethyl-, triester with boric acid h3bo3, tritrimethylsilyl borate, borane, tris trimethylsiloxy, silanol, 1,1,1-trimethyl-, 1,1',1-triester with boric acid h3bo3, acmc-209jtk, tris trimethylsiloxy borane, boric acid, 3tms derivative
PubChem CID 78020
IUPAC Name tris(trimethylsilyl) borate
SMILES B(O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Color Colorless
Density 0.8340g/mL
Boiling Point 183°C to 185°C
Flash Point 42°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula B[OSi(CH3)3]
Refractive Index 1.3850 to 1.387
Quantity 100 g
Specific Gravity 0.834
Formula Weight 278.38
Percent Purity ≥97.0%
Physical Form Liquid
Chemical Name or Material Tris(trimethylsilyl) borate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tris(trimethylsilyl) borate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.